Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery U-PLEX Proinflam Combo 1, multiplex immunoassay, 5 plate size, 2 plates provided, 96-well, detection limits sub- to low pg/mL, sample types: serum, plasma, cell culture supernatant
SDP

Supplier:  Meso Scale Discovery K15070K2

Encompass

The U-PLEX Proinflam Combo 1 (NHP) is a multiplex immunoassay designed for measuring nine key proinflammatory biomarkers in non-human primate samples. Utilizing advanced U-PLEX technology with electrochemiluminescence detection, this kit allows for precise cytokine and chemokine analysis across various sample types. It offers low detection limits and a broad dynamic range, making it an efficient solution for researchers. The kit includes two plates, enabling separation of incompatible assays, ensuring accurate performance for each biomarker.

  • Assay type: Multiplex immunoassay
  • Detection limits: sub- to low pg/mL range
  • Number of plates: 2
  • Plate type: 96-well
  • Compatible sample types: Serum, plasma, cell culture supernatant
  • Research use only: true
  • Storage condition: Multi-component

Catalog No. NC2209553


May include imposed supplier surcharges.
Add to Cart
Request a Quote
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.