Learn More
Meso Scale Discovery U-PLEX Proinflammatory Combo 1, 5 x 96-well plates, Human, Multiplex Immunoassay, ECL Detection, Serum, Plasma, Cell Culture Supernatant
SDP
Supplier:  Meso Scale Discovery K15049K2
Encompass
The MESO SCALE DISCOVERY U-PLEX Proinflammatory Combo 1 is a sophisticated multiplex immunoassay kit designed for the quantitative measurement of multiple human proinflammatory biomarkers. Leveraging MSD's MULTI-ARRAY technology, this kit offers remarkable sensitivity and a wide dynamic range, making it suitable for various high-throughput research applications in immunology and inflammation. The kit includes 5 plates, facilitating extensive testing with consistent results.
  • Analyte species includes human
  • Measures 10 specific proinflammatory biomarkers
  • Utilizes multiplex immunoassay assay type
  • Detection technology based on electrochemiluminescence
  • Configured in 96-well plate format
  • Kit size includes 5 plates
  • Compatible sample types: serum, plasma, and cell culture supernatant
  • Research use only
  • Supported by a range of MESO instruments
Catalog No. NC3442477
Add to Cart
Request a Quote