Learn More
Meso Scale Discovery U-PLEX Proinflam Combo 1 NHP, multiplex immunoassay, 96-well plate, serum, plasma, cell culture supernatant, detection range sub- to low pg/mL
SDP
Supplier:  Meso Scale Discovery K15070K1
Encompass
U-PLEX Proinflam Combo 1 NHP, multiplex immunoassay, 96-well plate, serum, plasma, cell culture supernatant, detection range sub- to low pg/mL
Catalog No. NC3405958