Learn More
Meso Scale Discovery U-PLEX Rat GLP-1 active ASY
SDP
Supplier:  Meso Scale Discovery K1536LK1
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay Kit is a multiplex immunoassay for measuring active GLP-1 in rat serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a 96-well plate with capture antibodies detection antibodies calibrator solutions diluent read buffer wash buffer and plate seals The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument It has a detection limit of 0 156 pg/mL for accurate GLP-1 quantification
Catalog No. 50-000-79758