Learn More
Meso Scale Discovery U-PLEX Rat Insulin ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K1536HK4
Encompass
The Meso Scale Discovery U-PLEX Rat Insulin Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of rat insulin in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive detection down to 0 05 ng/mL The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay Results are obtained in under 3 hours using an MSD instrument capable of reading electrochemiluminescent signals The assay is designed for research use only
Catalog No. 50-000-79368
Add to Cart
Request a Quote