Learn More
Meso Scale Discovery U-PLEX Rat Insulin ASY
SDP
Supplier:  Meso Scale Discovery K1536HK1
Encompass
The Meso Scale Discovery U-PLEX Rat Insulin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of rat insulin in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a lower limit of detection of 0 058 ng/mL and an upper limit of quantification of 1000 ng/mL It requires a low sample volume of 25 L and shows no cross-reactivity with mouse insulin The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and assay diluent
Catalog No. 50-000-82079