Learn More
Meso Scale Discovery U-PLEX Rat Insulin ASYQP
SDP
Supplier:  Meso Scale Discovery K1536HK22
Encompass
The Meso Scale Discovery U-PLEX Rat Insulin Assay Kit is a multiplex immunoassay that measures insulin levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluent The assay has a typical sensitivity of 0 058 pg/mL and an upper limit of quantification of 10 000 pg/mL It is for research use only
Catalog No. 50-000-80344