Learn More
Meso Scale Discovery U-PLEX Rat Insulin ASYQP
SDP
Supplier:  Meso Scale Discovery K1536HK21
Encompass
The Meso Scale Discovery U-PLEX Rat Insulin Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of rat insulin in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible detection of rat insulin on the QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and controls The assay has a typical lower limit of detection of 0 058 ng/mL and an upper limit of quantification of 1 000 ng/mL
Catalog No. 50-000-82181