Learn More
Meso Scale Discovery U-PLEX Rat Leptin ASY384-well
SDP
Supplier:  Meso Scale Discovery K2535ZK2
Encompass
The Meso Scale Discovery U-PLEX Rat Leptin 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of leptin in rat serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of samples The assay has a detection range of 50-100 000 pg/mL and includes a standard curve for quantitative analysis This product provides a sensitive reproducible method for measuring leptin levels in rat biological samples
Catalog No. 50-000-80009
Add to Cart
Request a Quote