Learn More
Meso Scale Discovery U-PLEX Rat Leptin ASYQP
SDP
Supplier:  Meso Scale Discovery K1535ZK21
Encompass
The Meso Scale Discovery U-PLEX Rat Leptin Assay Kit is a multiplex immunoassay for measuring leptin in rat serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags calibrators and controls The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument With a detection limit of 26 pg/mL it enables accurate leptin quantification for research
Catalog No. 50-000-81892