Learn More
Meso Scale Discovery U-PLEX TH1/TH2 Combo ms
SDP
Supplier:  Meso Scale Discovery K15071K2
Encompass
The U-PLEX TH1/TH2 Combo Mouse Assay is a multiplex immunoassay kit that allows simultaneous measurement of mouse IFN- IL-2 IL-4 IL-5 IL-10 and TNF- in cell culture supernatants and serum It uses electrochemiluminescence (ECL) detection technology and is compatible with the MESO QuickPlex SQ 120 instrument The assay requires a low sample volume of 25 L and has a short 3 5 hour protocol The kit includes a plate coated with capture antibodies detection antibodies calibrators controls wash buffer detection buffer read buffer sealing tape and plate sealers This assay allows efficient profiling of TH1 and TH2 immune responses in mouse samples
Catalog No. 50-000-78258
Only null left
Add to Cart
Request a Quote