Learn More
Meso Scale Discovery U-PLEX TH17 Combo 1 ms
SDP
Supplier:  Meso Scale Discovery K15077K2
Encompass
The U-PLEX TH17 Combo 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous quantitative measurement of IL-17A IL-17F IL-21 IL-22 IL-23 and IL-31 in mouse samples The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The electrochemiluminescent assay allows sensitive precise and accurate quantification of the 6 mouse Th17 pathway biomarkers from a single small sample volume The kit is for research use only
Catalog No. 50-000-79195
Only null left
Add to Cart
Request a Quote