Learn More
Meso Scale Discovery U-PLEX TH17 Combo 1 NHP
SDP
Supplier:  Meso Scale Discovery K15079K1
Encompass
The U-PLEX TH17 Combo 1 NHP is a multiplex assay kit by Meso Scale Diagnostics that enables simultaneous quantitative measurement of IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 IL-33 sCD40L and TNF It utilizes electrochemiluminescent detection technology and is designed for use with primate samples including human cynomolgus and rhesus This assay allows efficient profiling of multiple Th17-related biomarkers from a small sample volume With high sensitivity and broad dynamic range it is well-suited for research applications in immunology inflammation and autoimmunity The kit provides all necessary reagents and plates for analysis using Meso Scale Discovery instrumentation
Catalog No. 50-000-78280