Learn More
Meso Scale Discovery U-PLEX TH17 Combo 2, Kit, 10 biomarkers, Human, Electrochemiluminescence (ECL), Sub- to low pg/mL detection limit, 2 plates, Research Use Only, Multiplexing capability: Yes

Supplier: Meso Scale Discovery K15076K1
The MESO SCALE DISCOVERY U-PLEX TH17 Combo 2 kit is designed for the quantitative measurement of 10 biomarkers associated with the TH17 pathway in human samples. Utilizing advanced MULTI-ARRAY technology, this kit offers exceptional sensitivity and a broad dynamic range, making it a reliable choice for researchers. The kit features two plates to optimize separation of incompatible assays and is intended for research use only, ensuring optimal results for scientific investigations.
- Product type: Kit
- Target pathway: TH17
- Species: Human
- Number of biomarkers: 10
- Measurement type: Quantitative
- Detection method: Electrochemiluminescence (ECL)
- Detection limit: Sub- to low pg/mL
- Multiplexing capability: Yes
- Plate format: 2 plates
- Application: Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.