Learn More
Meso Scale Discovery U-PLEX TH17 Combo 2, multiplex immunoassay kit, 2 plates, 96-well, detects 10 cytokines, for serum, plasma, cell culture supernatant, storage: multi-component
SDP
Supplier:  Meso Scale Discovery K15076K2
Encompass
The U-PLEX TH17 Combo 2 (human) kit is designed for the quantitative measurement of 10 biomarkers related to the human Th17 pathway, ideal for research applications. This multiplex immunoassay kit allows for high sensitivity measurements from serum, plasma, and cell culture supernatant samples, with detection limits typically in the sub- to low pg/mL range. The assays are delivered on two 96-well plates, ensuring compatibility among all included analytes.
  • Multiplex compatible solution for biomarker measurement
  • High sensitivity with sub- to low pg/mL detection limits
  • Analyzes serum, plasma, and cell culture supernatant
  • Includes two 96-well plates
  • Detects 10 analytes associated with immune response
  • For research use only; not for diagnostic procedures
  • Multi-component storage requirements
  • Compatible with specified MESO instruments
  • Application in cytokines and chemokines studies
Catalog No. NC2456364
Only null left
Add to Cart
Request a Quote