Learn More
Meso Scale Discovery U-PLEX TH17 Combo 2 ms
SDP
Supplier:  Meso Scale Discovery K15078K1
Encompass
The U-PLEX TH17 Combo 2 Mouse Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous measurement of IL-17A IL-17F IL-21 IL-22 IL-23 and IL-31 in mouse samples such as serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology on patterned arrays for high sensitivity and broad dynamic range The assay requires 50 L of sample per well and can be read on MESO QuickPlex SQ 120 MESO SECTOR S 600 or MESO SECTOR S 6000 instruments It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance
Catalog No. 50-000-78279