Learn More
Ibidi USA µ-Slide 4 Well Ph+, ibiTreat - Tissue Culture Treated Polymer Coverslip
SDP
Supplier:  Ibidi USA 80446
Encompass
The ibidi product family is comprised of a variety of µ-Slides and µ-Dishes, which have all been designed for high-end microscopic analysis of fixed or living cells. The high optical quality of the polymer material is similar to that of glass. This allows the researcher to perform a wide variety of fluorescence experiments with uncompromised resolution and choice of wavelength. The µ-Slide 4 Well Ph+ ibiTreat is a convenient chambered coverslip with 4 wells for cell culture, immunofluorescence, and high-end microscopy. The µ-Slide 4 Well Ph+ improves the optical quality of phase contrast microscopy by suspending a plate in the center of the wells. When correctly filled, the plate eliminates the refraction of light due to meniscus formation compared to an open well. #1.5 polymer coverslip, tissue culture treated, sterilized, 15 pcs./box
Catalog No. NC0699518