Learn More
Montana Molecular G-GECO Calcium sensor Big Sky BacMam 5mL
SDP
Supplier:  Montana Molecular U0650G005
Encompass_Preferred
Detect Calcium mobilization or Gq signaling kinetics in living cells with the fluorescent G-GECO Calcium assay. Increase in green fluorescence indicates increase in Calcium. Packaged in Big Sky BacMam, for enhanced expression in mammalian cells including HEKs, CHOs, and many other cell types and iPSCs. This is an update to U0600G, with Big Sky BacMam providing improved expression efficiency.
Catalog No. 50-281-5250