Learn More
Medchemexpress LLC HY-107433 10mg Medchemexpress, U18666A CAS:3039-71-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10743310MG
Encompass_Preferred
Medchemexpress, HY-107433 10mg U18666A CAS:3039-71-2 U18666A, a cell permeable drug, is a cholesterol synthesis and transport inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8415