Learn More
Cayman Chemical U18666A Inhibitors
SDP
Supplier:  Cayman Chemical 100090855 MG
Encompass_Preferred
A cell permeable drug that inhibits cholesterol trafficking from late endosomes/lysosomes to the ER, but not to the plasma membrane
Catalog No. 50-289-0365