Learn More
Aviva Systems Biology Corp U1SNRNPBP Antibody - N-terminal region (ARP40699_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40699_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against U1SNRNPBP. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human U1SNRNPBP. Alternate names: HM-1, U1SNRNPBPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57593