Learn More
Eurofins Discoverx InCELL Hunter U2OS Bcl2L1-Bax Protein Binding Cell Line, 2 vials. (96-0009C3)
Supplier:  Eurofins Discoverx 960009C3
Encompass
The InCELL Hunter U2OS Bcl2L1-Bax Protein Binding Cell Line provides an easy-to-use cell based assay to study small molecule drug candidates that disrupt protein binding. This kit assesses compound-based disruption of Bcl2L1-Bax binding via detection of Bax levels. This product uses EFC technology. The cell line included overexpresses Bcl2L1 target in an untagged form and Bax binding partner fused to ProLabel. When a compound engages the Bcl2L1 target protein, partner binding is disrupted and the destabilized Bax is degraded, leading to a loss of EFC signal upon addition of EA containing detection reagent. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-4189
Add to Cart
Request a Quote