Learn More
Eurofins Discoverx PathHunter U2OS CCR10 β-Arrestin Cell Line, 2 vials. (93-0276C3)
Supplier:  Eurofins Discoverx 930276C3
Encompass
The PathHunter CCR10 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g.CCL27/CTACK ) based activation of CCR10 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCR10 and EA-tagged β-Arrestin-2. Activation of CCR10 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0150
Add to Cart
Request a Quote