Learn More
Eurofins Discoverx PathHunter U2OS CRTH2 β-Arrestin Cell Line, 2 vials. (93-0559C3)
Supplier:  Eurofins Discoverx 930559C3
Encompass
The PathHunter CRTH2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin D2) based activation of CRTH2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CRTH2 and EA-tagged β-Arrestin-2. Activation of CRTH2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0792
Add to Cart
Request a Quote