Learn More
Eurofins Discoverx PathHunter U2OS EphB4 Functional Assay, 2 vials. (93-0468C3)
Supplier:  Eurofins Discoverx 930468C3
Encompass
The PathHunter U2OS EphB4 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated EphB4 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged EphB4 and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-0591
Only null left
Add to Cart
Request a Quote