Learn More
Eurofins Discoverx PathHunter U2OS GPR17 β-Arrestin Cell Line, 2 vials. (93-0633C3)
Supplier:  Eurofins Discoverx 930633C3
Encompass
The PathHunter GPR17 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. MDL 29,951) based activation of GPR17 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR17 and EA-tagged β-Arrestin-2. Activation of GPR17 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0984
Add to Cart
Request a Quote