Learn More
Eurofins Discoverx PathHunter U2OS IL1RL1/IL1RAP Dimerization Cell Line, 2 vials. (93-1067C3)
Supplier:  Eurofins Discoverx 931067C3
Encompass
The PathHunter IL1RL1/IL1RAP Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-33) induced dimerization of IL1RL1/IL1RAP receptors. This product uses EFC technology. The cell line included overexpresses IL1RL1 target protein fused to PK alongside IL1RAP target protein fused to EA. Upon ligand-induced dimerization of IL1RL1/IL1RAP, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1967
Only null left
Add to Cart
Request a Quote