Learn More
Eurofins Discoverx PathHunter U2OS IL23R/IL12RB1 Dimerization Cell Line, 2 vials. (93-1007C3)
Supplier:  Eurofins Discoverx 931007C3
Encompass
The PathHunter IL23R/IL12RB1 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-23) induced dimerization of IL23R/IL12RB1 receptors. This product uses EFC technology. The cell line included overexpresses IL23R target protein fused to PK alongside IL12RB1 target protein fused to EA. Upon ligand-induced dimerization of IL23R/IL12RB1, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1823
Only null left
Add to Cart
Request a Quote