Learn More
Eurofins Discoverx PathHunter U2OS mBRS3 β-Arrestin Cell Pool, 2 vials. (93-0597P3)
Supplier:  Eurofins Discoverx 930597P3
Encompass
The PathHunter mBRS3 cell pool is a stable, engineered cell pool for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. bombesin) based activation of mBRS3 GPCR activity via detection of β-arrestin recruitment. The included cell pool overexpresses ProLink-tagged mBRS3 and EA-tagged β-Arrestin-2. Activation of mBRS3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0919
Add to Cart
Request a Quote