Learn More
Eurofins Discoverx PathHunter U2OS MC5R β-Arrestin Cell Line, 2 vials. (93-0231C3)
Supplier:  Eurofins Discoverx 930231C3
Encompass
The PathHunter MC5R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. α-MSH) based activation of MC5R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged MC5R and EA-tagged β-Arrestin-2. Activation of MC5R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0011
Add to Cart
Request a Quote