Learn More
Eurofins Discoverx PathHunter U2OS mMC5R β-Arrestin Cell Pool, 2 vials. (93-0609P3)
Supplier:  Eurofins Discoverx 930609P3
Encompass
The PathHunter mMC5R cell pool is a stable, engineered cell pool for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. melanotan II) based activation of mMC5R GPCR activity via detection of β-arrestin recruitment. The included cell pool overexpresses ProLink-tagged mMC5R and EA-tagged β-Arrestin-2. Activation of mMC5R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0937
Only null left
Add to Cart
Request a Quote