Learn More
Eurofins Discoverx PathHunter U2OS mP2RY12 β-Arrestin Cell Line, 2 vials. (93-0695C3)
Supplier:  Eurofins Discoverx 930695C3
Encompass
The PathHunter mP2RY6 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. UTP) based activation of mP2RY6 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged mP2RY6 and EA-tagged β-Arrestin-2. Activation of mP2RY6 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1134
Add to Cart
Request a Quote