Learn More
Eurofins Discoverx PathHunter U2OS PRLR-JAK2 Functional Assay, 2 vials. (93-0687C3)
Supplier:  Eurofins Discoverx 930687C3
Encompass
The PathHunter PRLR-JAK2 Functional assay cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting CTKs. This cell line enables assessment of PRLR activation via detection of JAK2 mediated phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged PRLR cytokine receptor, an untagged CTK JAK2 and EA-tagged SH2 domain protein. Upon activation of the cytokine receptor, CTK-based receptor phosphorylation leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1107
Only null left
Add to Cart
Request a Quote