Learn More
Eurofins Discoverx PathHunter U2OS PTGFR β-Arrestin Cell Line, 2 vials. (93-0936C3)
Supplier:  Eurofins Discoverx 930936C3
Encompass
The PathHunter PTGFR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. cloprostenol) based activation of PTGFR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTGFR and EA-tagged β-Arrestin-2. Activation of PTGFR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1605
Only null left
Add to Cart
Request a Quote