Learn More
Eurofins Discoverx PathHunter U2OS SMO(W535L) MEM-EA Pharmacotrafficking Assay, 2 vials. (93-0991C3)
Supplier:  Eurofins Discoverx 930991C3
Encompass
The PathHunter U2OS SMO(W535L) MEM-EA Pharmacotrafficking Assay is a stable, engineered cell line for use in studying pharmacochaperones targeting SMO. This cell line enables assessment of chaperone-based stabilization of SMO(W535L) via detection of rescued membrane trafficking. The included cell line overexpresses ProLink-tagged mutant SMO(W535L), which is misfolded and retained in the ER. The cell line also expresses membrane-localized EA. When a pharmacochaperone stabilizes the mutant SMO, it is trafficked to the membrane, producing EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1763
Add to Cart
Request a Quote