Learn More
Eurofins Discoverx PathHunter U2OS TORC3 Nuclear Translocation Cell Line, 2 vials. (93-1120C3)
Supplier:  Eurofins Discoverx 931120C3
Encompass
The PathHunter U2OS TORC3 Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the CRTC/CREB pathway. This cell line enables assessment of ligand (e.g. forskolin) based pathway activation via detection of resulting TORC3 (CRTC3) nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged TORC3 (CRTC3) and nuclear localized EA. Upon pathway activation, induced nuclear translocation of TORC3 leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.
Catalog No. 50-200-2140
Add to Cart
Request a Quote