Learn More
Selleck Chemical LLC U73122
SDP
Supplier:  Selleck Chemical LLC S80111G
Encompass_Preferred
U73122 is a potent phospholipase C (PLC) inhibitor which reduces agonist-induced Ca2 increases in platelets and PMN U73122 potently inhibits human 5-lipoxygenase (5-LO)
Catalog No. 50-000-00870