Learn More
ATCC UACC-3133 (ATCC CRL-2988) Human breast cancer cell line - infiltrating lobular carcinoma metastatic to pleural fluid
Supplier: ATCC CRL2988
This human breast cancer cell line, UACC-3133 (CRL-2988), was derived from pleural fluid of a 52-year-old female with poorly differentiated adenocarcinoma consistent with infiltrating lobular carcinoma. The line exhibits epithelial morphology, adherent growth, and a distinct marker profile useful for breast cancer research.
- Epithelial-like cell morphology.
- Adherent growth suitable for monolayer and 3D culture.
- Neu (ErbB2/HER2) positive; low estrogen receptor expression.
- Progesterone receptor and EGFR negative.
- High BMP3 expression with lack of BMP2.
- Recommended medium: Leibovitz's L-15 with serum and supplements.
- Culture conditions: 37°C, 100% air; subculture 1:2; renew medium every 3-4 days.
- Mycoplasma not detected; store in vapor phase of liquid nitrogen.