Learn More
Medchemexpress LLC UAMC-3203 hydrochloride | 2271358-65-5 | 98.8% | 508.12 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY112909A10MM/1ML
Encompass_Preferred
UAMC-3203 hydrochloride is a potent and selective Ferroptosis inhibitor with an IC50 of 12 nM. No toxicity was observed in mice after repeated intraperitoneal injections (20 μmol/kg daily, over a period of 4 weeks).
  • Potent and selective ferroptosis inhibitor.
  • Has an IC50 of 12 nM for ferroptosis.
  • No observed toxicity in mice at 20 μmol/kg daily for 4 weeks.
  • Available in solid form (white to off-white).
  • High purity: 98.82%.
Catalog No. 50-004-40032