Learn More
Ubpbio A commonly used fluorogenic substrate for deubiquitinating enzymes (DUBs)
Supplier:  Ubpbio M3022
Encompass_Preferred
Ub-Rhodamine 110 is a quenched, fluorescent substrate for deubiquitinating enzymes. Cleavage of the amide bond between the C-terminal glycine of ubiquitin and rhodamine110 results in an increase in rhodamine fluorescence at 535 nm (Exc. 485 nm).
Catalog No. 50-233-9257