Learn More
LifeSensors UBE2T 3nmol
SDP
Supplier:  LifeSensors UB02190030
Encompass_Preferred
UBE2T can mediate the transfer of ubiquitin from ubiquitin-activating enzyme E1 to a substrate protein or E3 ligase. This E2 enzyme has been shown to be essential for the Fanconi anemia pathway. UBE2T conjugating activity is involved with the monoubiquitylation of FANCD2, co-functioning with the E3 ligase FANCL.
Catalog No. NC3354980