Learn More
ABclonal Technology UBA5 Rabbit pAb
SDP
Supplier:  ABclonal Technology A155141000UL
Encompass_Preferred
This gene encodes a member of the E1-like ubiquitin-activating enzyme family. This protein activates ubiquitin-fold modifier 1, a ubiquitin-like post-translational modifier protein, via the formation of a high-energy thioester bond. Alternative splicing results in multiple transcript variants. A pseudogene of this gene has been identified on chromosome 1.
Catalog No. 50-001-05579