Learn More
Selleck Chemical LLC UBCS039 5mg
SDP
Supplier:  Selleck Chemical LLC S89095MG
Encompass_Preferred
UBCS039 is the first synthetic SIRT6 activator with EC50 of 38 ?M,induces a time-dependent activation of autophagy in several human tumor cell lines. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3428