Learn More
Enzo Life Sciences Ubiquitinylation kit (1x1Kit)
SDP
Supplier:  Enzo Life Sciences BMLUW99200001
Encompass_Preferred
Versatile tool for generation of ubiquitin-E2 thioesters. Contents: 20X Ubiquitin Activating Enzyme Solution Human recombinant E1 (His6-tagged), 125µl provided, Product No. BML-UW9410 10X Ubiquitin Conjugating Enzyme Solutions (E2) 20µl of each E2 provided UbcH1 (His6-tagged), Prod. No. BML-UW9020 UbcH2 (His6-tagged), Prod. No. BML-UW9025 UbcH3 (His6-tagged), Prod. No. BML-UW8730 UbcH5a (His6-tagged), Prod. No. BML-UW9050 UbcH5b (His6-tagged), Prod. No. BML-UW9060 UbcH5c (His6-tagged), Prod. No. BML-UW9070 UbcH6 (His6-tagged), Prod. No. BML-UW8710 UbcH7 (His6-tagged), Prod. No. BML-UW9080 UbcH8 (His6-tagged), Prod. No. BML-UW9135 UbcH10 (untagged), Prod. No. BML-UW0960 UbcH13/Mms2 (His6-tagged), Prod. No. BML-UW9565 20X Biotinylated Ubiquitin Solution (Bt-Ub), 125µl provided, Prod. No. BML-UW8705 20X Mg-ATP Solution, 125µl provided, Prod. No. BML-EW9805 2X Non-reducing Gel Loading Buffer, 2.5 ml, Prod. No. BML-KW9880 10X Ubiquitinylation Buffer, 250 µl, Prod. No. BML-KW9885
Catalog No. 50-103-1803