Learn More
Waters Corp Waters μBondapak C18 Column, 125Å, 10 μm, 3.9 mm × 300 mm
Supplier:  Waters Corp WAT027324
Encompass_Preferred
μBondapak C18 columns are silica-based, reversed-phase C18 columns utilizing 10 μm particle technology, designed for preparative chromatography with a balance of resolution and throughput.
  • Silica-based, reversed-phase C18 column with 10 μm particle technology.
  • Provides optimal balance of resolution, throughput, and cost for preparative chromatography.
  • Offers unique selectivity due to controlled hydrophobicity and silanol activity.
  • Manufactured in an ISO 9002-certified facility with stringent quality control.
  • Ideal for analyzing hydrophobic and moderately polar compounds, including small molecules and weakly basic analytes.
  • Compatible with a wide range of water-organic solvent mobile phases for pH control.
  • Delivers high efficiency and superior peaks; suitable for scale-up and new purification protocols.
Catalog No. 50-818-611