Learn More
Waters Corp Waters μBondapak Cartridge Column, C18, 10 μm, 4.6 mm X 250 mm
Supplier:  Waters Corp WAT052860
Encompass_Preferred
μBondapak C18 columns are silica-based, reversed-phase columns utilizing 10 μm particle technology. They provide a balance of resolution, throughput, and cost for preparative chromatography. Known for year-to-year reproducibility, these columns are manufactured with strict quality control to ensure consistent results in chromatographic applications, especially for methods requiring irregular and non-spherical materials.
  • Silica-based, reversed-phase C18 columns with 10 μm particle technology.
  • Optimal balance of resolution, throughput, and cost for preparative chromatography.
  • Known for excellent year-to-year reproducibility.
  • Manufactured under strict quality control processes.
  • Ensures high quality and reproducible chromatographic results.
  • Supports methods requiring irregular and non-spherical materials.
Catalog No. 50-785-686