Learn More
Apexbio Technology LLC UBP1112 339526-74-8 50mg
SDP
Supplier:  Apexbio Technology LLC B670650
Encompass_Preferred
UBP1112 is an antagonist targeting type III metabotropic glutamate (mGlu) receptors a subgroup of G protein-coupled receptors involved in synaptic modulation of excitatory neurotransmission within the central nervous system Structurally UBP1112 interacts specifically with these receptors blocking glutamate binding and subsequently altering downstream signal transduction In neuroscience research UBP1112 serves as an investigative tool for examining receptor-mediated neuronal signaling pathways and for studying pathophysiological mechanisms underlying neurodegenerative conditions
Catalog No. 50-000-62871