Learn More
Medchemexpress LLC UBP608 | 2199-87-3 | MFCD00047640 | 99.6% | 269.05 g/mol | C10H5BrO4 | 10 G
SDP
Supplier:  Medchemexpress LLC HY10066710G
Encompass_Preferred
UBP608 is a small-molecule negative allosteric modulator of N-methyl-D-aspartate receptors (NMDARs) used in preclinical neuroscience research. It is supplied as a high-purity research reagent suitable for studies of NMDAR function and models of neurological disorders.
  • Potent negative allosteric modulator of N-methyl-D-aspartate receptors.
  • High purity, 99.6%.
  • Molecular formula C10H5BrO4 and molecular weight 269.05 g/mol.
  • Registered CAS number 2199-87-3 for unambiguous identification.
  • Stable as a powder under recommended low-temperature storage conditions.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
Catalog No. 50-002-13710