Learn More
Medchemexpress LLC UC-764864 | 278806-03-4 | C19H18N2OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1537755MG
Encompass_Preferred
UC-764864 is a UBE2N inhibitor that impedes UBE2N enzymatic activity. It exhibits cytotoxic effects and hinders UBE2N-dependent signaling in leukemic cells. Furthermore, UC-764864 blocks the ubiquitination of innate immune- and inflammatory-related substrates within human Acute Myeloid Leukemia (AML) cell lines.
  • Inhibits UBE2N enzymatic activity
  • Demonstrates cytotoxic effects in leukemic cells
  • Inhibits UBE2N-dependent signaling in leukemic cells
  • Blocks ubiquitination of innate immune- and inflammatory-related substrates in human AML cell lines
Catalog No. 50-003-76306