Learn More
Medchemexpress LLC UCB-35440 | 299460-62-1 | 99.4% | 576.09 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY194431MG
Encompass_Preferred
UCB-35440 is an orally active histamine H1 receptor antagonist and 5-lipoxygenase inhibitor. It inhibits LTB4 formation in human whole blood and reduces polymorphonuclear cell infiltration in mouse models. It inhibits histamine-stimulated bronchoconstriction and reduces skin inflammation in guinea pigs, and can be used for research on asthma and skin inflammation.
  • Orally active histamine H1 receptor antagonist and 5-lipoxygenase inhibitor.
  • Inhibits LTB4 formation in human whole blood.
  • Reduces polymorphonuclear cell infiltration in mouse models.
  • Inhibits histamine-stimulated bronchoconstriction.
  • Reduces skin inflammation in guinea pigs.
  • Can be used for research on asthma and skin inflammation.
Catalog No. 50-003-85078